Stampa…

Ethyl benzimidate hydrochloride 97%

Fornitore: Thermo Scientific
Attenzione

L17684.09 L17684.18
L17684.09EA 34.4 EUR
L17684.09 L17684.18
Ethyl benzimidate hydrochloride 97%
Ethyl benzimidate hydrochloride

Purity: 97% CAS No.: 5333-86-8 Molecular Formula: C9H11NO.HCl Formula Weight: 185.65 Melting Point: 115-117° Beilstein Reference: 3697589 MDL No.: MFCD00043246

Formula: C₉H₁₂ClNO
Punto di fusione: 115…117 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00043246
CAS Number: 5333-86-8
EINECS: 226-248-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR