Stampa…

Pyromellitic diimide 97%

Fornitore: Thermo Scientific

Synonyms: Benzene-1,2,4,5-tetracarboxylic Diimide

L14372.06 L14372.14
L14372.06EA 38.7 EUR
L14372.06 L14372.14
Pyromellitic diimide 97%
Pyromellitic diimide

Purity: 97% CAS No.: 2550-73-4 Molecular Formula: C10H4N2O4 Formula Weight: 216.15 Melting Point: 300° Beilstein Reference: 214780 MDL No.: MFCD00005004

Formula: C₁₀H₄N₂O₄
Peso molecolare: 216,15 g/mol
Punto di fusione: >300 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005004
CAS Number: 2550-73-4
EINECS: 219-852-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR