N,N'-Ethylenebisacrylamide 96%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 96%
CAS No.: 2956-58-3
Molecular Formula: C8H12N2O2
Formula Weight: 168.20
Storage Temperature: 2°C to 8°C
Melting Point: 141-143°
Beilstein Reference: 1771570
MDL No.: MFCD00042935
Formula:
C₈H₁₂N₂O₂ Punto di fusione: 141…143 °C |
MDL Number:
MFCD00042935 CAS Number: 2956-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...