Stampa…

N-Benzylacrylamide 96%

Fornitore: Thermo Fisher Scientific
Attenzione

L11450.03 L11450.06
L11450.03EA 39.9 EUR
L11450.03 L11450.06
N-Benzylacrylamide 96%
N-Benzylacrylamide

Purity: 96% CAS No.: 13304-62-6 Molecular Formula: CH2=CHCONHCH2C6H5 Formula Weight: 161.21 Melting Point: 59-62° Beilstein Reference: 2690053 MDL No.: MFCD00015333

Formula: CH₂=CHCONHCH₂C₆H₅
Punto di fusione: 59…62 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00015333
CAS Number: 13304-62-6
EINECS: 603-701-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR