Methylhydrazine sulphate 98%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Purity: 98%
CAS No.: 302-15-8
Molecular Formula: CH3NHNH2.H2SO4
Formula Weight: 144.15
Melting Point: 141-143°
Beilstein Reference: 3693575
UN No.: UN2811
MDL No.: MFCD00035423
Formula:
CH₆N₂ Punto di fusione: 141…143 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00035423 CAS Number: 302-15-8 EINECS: 206-115-4 UN: 2811 ADR: 6.1,III Merck Index: 14,6086 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...