3,5-Bis(trifluoromethyl)phenyl Isocyanate 98%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Purity: 98%
CAS No.: 16588-74-2
Molecular Formula: C9H3F6NO
Formula Weight: 255.12
Storage Temperature: 2°C to 8°C
Flash Point: 67°(152°F)
Refractive Index: 1.4300
Odour: Pungent
Boiling Point: 62°/2mm
Beilstein Reference: 542272
UN No.: UN2206
MDL No.: MFCD00013559
Formula:
C₉H₃F₆NO Punto di ebollizione: 62 °C (2 mmHg) Densità: 1,476 Flash Pt: 67 °C (152 °F) |
MDL Number:
MFCD00013559 CAS Number: 16588-74-2 ADR: 6.1,III |