Stampa…

1-Hexen-3-one ≥90% stabilizzato

Fornitore: Thermo Scientific
Pericolo


Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 0,5%

L07660.06 L07660.14
L07660.06EA 118 EUR
L07660.06 L07660.14
1-Hexen-3-one ≥90% stabilizzato
1-Hexen-3-one

Stab. with 0.5% 4-methoxyphenol


Purity: 90+% CAS No.: 1629-60-3 Molecular Formula: C6H10O Formula Weight: 98.15 Storage Temperature: 2°C to 8°C Flash Point: 22°(71°F) Refractive Index: 1.4275 Odour: Pungent Boiling Point: 127-129° Beilstein Reference: 1740796 UN No.: UN1224 MDL No.: MFCD00051563

Formula: C₆H₁₀O
Peso molecolare: 98,15 g/mol
Punto di ebollizione: 127…129 °C
Densità: 0,840
Flash Pt: 22 °C (71 °F)
MDL Number: MFCD00051563
CAS Number: 1629-60-3
EINECS: 216-625-9
ADR: 3,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR