4-Ethoxycinnamic acid predominantly trans ≥98%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
3-(4-Ethoxyphenyl)acrylic Acid
L06324.06
L06324.14
L06324.06EA
77.5
EUR
L06324.06
L06324.14
4-Ethoxycinnamic acid predominantly trans ≥98%
4-Ethoxycinnamic acid
Purity: 98+%
CAS No.: 2373-79-7
Molecular Formula: C11H12O3
Formula Weight: 192.22
Melting Point: 195-199°
Beilstein Reference: 3198068
MDL No.: MFCD00016848
Formula:
C₁₁H₁₂O₃ Peso molecolare: 192,21 g/mol Punto di fusione: 195…199 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11040581 CAS Number: 2373-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...