N-(2-Chloroethyl)benzamide 97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 97%
CAS No.: 26385-07-9
Molecular Formula: C9H10ClNO
Formula Weight: 183.64
Melting Point: 101-105°
Beilstein Reference: 1943939
MDL No.: MFCD00000964
Formula:
C₉H₁₀ClNO Peso molecolare: 183.635 g/mol Punto di fusione: 101…105 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00000964 CAS Number: 26385-07-9 EINECS: 247-641-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...