Methyl-4-(methylsulphonylmethyl)benzoate 98%
Fornitore: Thermo Scientific
L05657.04
L05657.09
L05657.04EA
71.3
EUR
L05657.04
L05657.09
Methyl-4-(methylsulphonylmethyl)benzoate 98%
Methyl-4-(methylsulphonylmethyl)benzoate
Purity: 98%
CAS No.: 160446-22-0
Molecular Formula: C10H12O4S
Formula Weight: 228.27
Melting Point: 164-166°
Beilstein Reference: 7810971
MDL No.: MFCD00051693
Formula:
C₁₀H₁₂O₄S Peso molecolare: 228,27 g/mol Punto di fusione: 164…166 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00051693 CAS Number: 160446-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...