Ethyl-2-pentynoate ≥97%
Fornitore: Thermo Scientific
Attenzione
|
Synonyms:
Ethyl pent-2-ynoate
Purity: 98%
CAS No.: 55314-57-3
Molecular Formula: C7H10O2
Formula Weight: 126.16
Flash Point: 64°(147°F)
Refractive Index: 1.4395
Boiling Point: 178-179°
Beilstein Reference: 1750016
MDL No.: MFCD00015221
Formula:
C₇H₁₀O₂ Peso molecolare: 126,155 g/mol Punto di ebollizione: 178…179 °C Densità: 0,957 Flash Pt: 64 °C (147 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00015221 CAS Number: 55314-57-3 EINECS: 259-588-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...