1-Adamantanecarbonyl chloride 97%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
1-Adamantanecarboxylic acid chloride
Purity: 97%
CAS No.: 2094-72-6
Molecular Formula: C11H15ClO
Formula Weight: 198.69
Melting Point: 48-53°
Boiling Point: 135-136°/10mm
Beilstein Reference: 389960
UN No.: UN3261
MDL No.: MFCD00074724
Formula:
C₁₁H₁₅ClO Peso molecolare: 198,69 g/mol Punto di ebollizione: 135…136 °C (10 mmHg) Punto di fusione: 48…53 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00074724 CAS Number: 2094-72-6 EINECS: 218-252-7 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...