Stampa…

5-Hydroxymethyluracil 98%

Fornitore: Thermo Scientific

Synonyms: 5-(Hydroxymethyl)pyrimidine-2,4(1H,3H)-dione

L01682.03 L01682.06
L01682.03EA 50.2 EUR
L01682.03 L01682.06
5-Hydroxymethyluracil 98%
5-Hydroxymethyluracil

Purity: 98% CAS No.: 4433-40-3 Molecular Formula: C5H6N2O3 Formula Weight: 142.12 Melting Point: 300° Beilstein Reference: 125482 MDL No.: MFCD00006070

Formula: C₅H₆N₂O₃
Peso molecolare: 142,114 g/mol
Punto di fusione: >300 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006070
CAS Number: 4433-40-3
EINECS: 224-636-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR