Stampa…

Oro 95% supplied in 0.1 mM PBS, nanoparticles 150 nm

Fornitore: Thermo Fisher Scientific

J67276.ACEA 110 EUR
J67276.AC J67276.AE
Oro 95% supplied in 0.1 mM PBS, nanoparticles 150 nm
Oro

Purity: 95% CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Dark brown to pink MDL No.: MFCD00003436


Applications: For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

Formula: Au
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR