Crocein scarlet 7B ≥70% (dye content)
Fornitore: Thermo Fisher Scientific
CAS No.: 6226-76-2
Molecular Formula: C24H18N4Na2O7S2
Formula Weight: 584.53
Physical Form: Powder
Appearance: Red
Melting Point: 181-188°
Solubility: Soluble in water
MDL No.: MFCD00056588
Formula:
C₂₄H₁₈N₄Na₂O₇S₂ Punto di fusione: 181…188 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00056588 CAS Number: 6226-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...