Ethylenediamine-N,N'-diacetic acid
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
Ethylene-N,N'-diglycine, EDDA
CAS No.: 5657-17-0
Molecular Formula: C6H12N2O4
Formula Weight: 176.17
Physical Form: Powder
Appearance: White
Melting Point: 224°
Beilstein Reference: 1778355
MDL No.: MFCD00004281
Applications: Acts as a chelating agent
Formula:
C₆H₁₂N₂O₄ Peso molecolare: 176,17 g/mol Punto di fusione: 224 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00004281 CAS Number: 5657-17-0 EINECS: 227-105-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...