Stampa…

Enalaprilat

Fornitore: Thermo Scientific

Synonyms: (S)-1-((S)-2-(((S)-1-Carboxy-3-phenylpropyl)amino)propanoyl)pyrrolidine-2-carboxylic acid

J63392.MA J63392.MF
J63392.MAEA 144 EUR
J63392.MA J63392.MF
Enalaprilat
Enalaprilat

CAS No.: 76420-72-9 Molecular Formula: C18H28N2O7 Formula Weight: 384.42 Physical Form: Crystalline solid Appearance: White Melting Point: 148-151° Solubility: Soluble in water with heating (~60°C) MDL No.: MFCD00865786


Applications: The active metabolite of enalapril

Formula: C₁₈H₂₈N₂O₇
Peso molecolare: 384,42 g/mol
Punto di fusione: 148…151 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00865786
CAS Number: 76420-72-9
EINECS: 278-459-3
Merck Index: 14,3568

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR