Paclitaxel ≥99.5%
Fornitore: Thermo Scientific
|
Pericolo
|
Purity: 99.5+%
CAS No.: 33069-62-4
Molecular Formula: C47H51NO14
Formula Weight: 853.91
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline solid
Appearance: White
Melting Point: 213° dec.
Solubility: Soluble in DMSO (200mg/ml) or ethanol (40mg/ml)
Beilstein Reference: 1420457
MDL No.: MFCD00869953
Applications: Promotes assembly and inhibits disassembly of microtubules (anti-cancer agent)
Formula:
C₄₇H₅₁NO₁₄ Peso molecolare: 853,91 g/mol Punto di fusione: 213 °C (dec.) |
MDL Number:
MFCD00869953 CAS Number: 33069-62-4 Merck Index: 14,6982 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...