Stampa…

Triolein ≥75%, tech.

Fornitore: Thermo Scientific

Synonyms: Glyceryl trioleate, Glycerine trioleate, Glycerol Trioleate

J62419.14 J62419.22
J62419.14EA 165 EUR
J62419.14 J62419.22
Triolein ≥75%, tech.
Triolein

Purity: 75% CAS No.: 122-32-7 Molecular Formula: C57H104O6 Formula Weight: 885.45 Storage Temperature: -30°C to -10°C Flash Point: 330°(626°F) Physical Form: Liquid Density: 0.91 Refractive Index: 1.468 Appearance: Clear pale yellow Boiling Point: 235-240°/18mm Solubility: Soluble in chlorofom, ether, and carbon tetrachloride Beilstein Reference: 1718692 MDL No.: MFCD00137563

Formula: C₅₇H₁₀₄O₆
Peso molecolare: 885,45 g/mol
Punto di ebollizione: 235…240 °C (18 mmHg)
Densità: 0,91
Flash Pt: 330 °C (626 °F)
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00137563
CAS Number: 122-32-7
EINECS: 204-534-7
Merck Index: 14,9732

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR