Minoxidil
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Synonyms:
2,6-Diamino-4-(piperidin-1-yl)pyrimidine 1-oxide, Rogaine, Loniten
CAS No.: 38304-91-5
Molecular Formula: C9H15N5O
Formula Weight: 209.25
Physical Form: Crystalline powder
Appearance: White
Melting Point: 248-265°
MDL No.: MFCD00063409
Applications: Stimulates ATP-activated potassium channels
Formula:
C₉H₁₅N₅O Punto di fusione: 248…265 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00063409 CAS Number: 38304-91-5 EINECS: 253-874-2 Merck Index: 14,6203 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...