Stampa…

Erioglaucine disodium salt

Fornitore: Thermo Fisher Scientific

Synonyms: FD and C Blue No. 1, Brilliant Blue FCF, Erioglaucin A, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulphonate, Alphazurine FG, Erioglaucine sodium salt, CI 42090, Acid blue 9, Food Blue No.1

J61780.06 J61780.14
J61780.06EA 29.6 EUR
J61780.06 J61780.14
Erioglaucine disodium salt
Erioglaucine disodium salt

CAS No.: 3844-45-9 Molecular Formula: C37H34N2Na2O9S3 Formula Weight: 792.86 Physical Form: Powder Appearance: Purple Melting Point: 283° MDL No.: MFCD00012141

Formula: C₃₇H₃₄Na₂N₂O₉S₃
Punto di fusione: 283 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012141
CAS Number: 3844-45-9
EINECS: 223-339-8
Merck Index: 14,1373

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR