Stampa…

Harmaline ≥98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 3,4-Dihydro-7-methoxy-1-methyl-beta-carboline, 3,4-Dihydro-7-methoxy-1-methyl-9H-pyrido[3,4-b]indole, Dihydroharmine

J61699.03 J61699.06
J61699.03EA 57.4 EUR
J61699.03 J61699.06
Harmaline ≥98%
Harmaline

Purity: 98+% CAS No.: 304-21-2 Molecular Formula: C13H14N2O Formula Weight: 214.27 Storage Temperature: 2°C to 8°C Physical Form: Crystals Appearance: Pale brown Melting Point: 239-241° Beilstein Reference: 207310 MDL No.: MFCD00004955


Applications: CNS stimulant; may act through NMDA receptors. Reversible inhibitor of MAO-A

Formula: C₁₃H₁₄N₂O
Peso molecolare: 214,27 g/mol
Punto di fusione: 239…241 °C
MDL Number: MFCD00004955
CAS Number: 304-21-2
EINECS: 206-152-6
Merck Index: 14,4613

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR