Lincomycin hydrochloride
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
CAS No.: 859-18-7
Molecular Formula: C18H34N2O6S.HCl
Formula Weight: 443.00
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White to off-white
Solubility: Soluble in water
Beilstein Reference: 4171650
MDL No.: MFCD00058237
Applications: Lincosamide anitbiotic active against Gram-positive bacteria
Formula:
C₁₈H₃₄N₂O₆S·HCl Peso molecolare: 443.0 g/mol (anhy) |
MDL Number:
MFCD00058237 CAS Number: 859-18-7 EINECS: 212-726-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...