Ribonucleic acid (from bakers yeast)
Fornitore: Thermo Fisher Scientific
Ribonucleic acid (RNA) from baker's yeast S. cerevisiae may be used as a substrate for studying ribonuclease activities of enzymes such as RNAase(s). It is also suitable for use as a carrier in nucleic acid purification and precipitation.
- Off-white to pale brown powder
- Soluble in water
- Store at cold temperature
Suitable for use as a ribonuclease substrate.
Temperatura di stoccaggio:
Refrigerata |
MDL Number:
MFCD00132195 CAS Number: 63231-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...