Tricine-buffered saline (5X), pH 8,0
Fornitore: Thermo Scientific
J60745.AK
J60745.AP
J60745.AKEA
58.1
EUR
J60745.AK
J60745.AP
Tricine-buffered saline (5X), pH 8,0
Buffers
Tamponi biologici
CAS No.: 5704-04-1
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00004277
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...