(R)-(+)-1-(1-Naphthyl)ethylamine 97%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
(R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine
Purity: 97%
CAS No.: 3886-70-2
Molecular Formula: C12H13N
Formula Weight: 171.24
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₂H₁₃N Peso molecolare: 171,24 g/mol Punto di ebollizione: 152…153 °C (11 mmHg) Densità: 1,060 Flash Pt: >110 °C (230 °F) |
MDL Number:
MFCD00064114 CAS Number: 3886-70-2 EINECS: 223-425-5 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...