Stampa…

5-Nitro-2-thiophenecarboxylic acid 98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 5-Nitrothiophene-2-carboxylic acid

H64821.06 H64821.14 H64821.22
H64821.06EA 56 EUR
H64821.06 H64821.14 H64821.22
5-Nitro-2-thiophenecarboxylic acid 98%
5-Nitro-2-thiophenecarboxylic acid

Purity: 98% CAS No.: 6317-37-9 Molecular Formula: C5H3NO4S Formula Weight: 173.14 Physical Form: Solid Appearance: Pale yellow Melting Point: 154-159° Beilstein Reference: 139665 MDL No.: MFCD00159552

Formula: C₅H₃NO₄S
Peso molecolare: 173,14 g/mol
Punto di fusione: 154…159 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00159552
CAS Number: 6317-37-9
EINECS: 228-654-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR