9-Bromo-10-phenylanthracene 98%
Fornitore: Thermo Scientific
|
Attenzione
|
H64794.03
H64794.06
H64794.03EA
70.8
EUR
H64794.03
H64794.06
9-Bromo-10-phenylanthracene 98%
9-Bromo-10-phenylanthracene
Purity: 98%
CAS No.: 23674-20-6
Molecular Formula: C20H13Br
Formula Weight: 333.23
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 154-158°
Beilstein Reference: 2277064
MDL No.: MFCD00230983
Formula:
C₂₀H₁₃Br Peso molecolare: 333,23 g/mol Punto di fusione: 154…158 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00230983 CAS Number: 23674-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...