(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 97%
CAS No.: 869681-70-9
Molecular Formula: C10H17NO5
Formula Weight: 231.25
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 181°
Beilstein Reference: 10254221
UN No.: UN2811
MDL No.: MFCD04114909
Formula:
C₁₀H₁₇NO₅ Punto di fusione: 181 °C |
MDL Number:
MFCD04114909 CAS Number: 869681-70-9 ADR: 6.1,III |