Stampa…

2-Bromodibenzothiophene 98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 2-Bromodibenzo[b,d]thiophene

H64645.06 H64645.14
H64645.06EA 146 EUR
H64645.06 H64645.14
2-Bromodibenzothiophene 98%
2-Bromodibenzothiophene

Purity: 98% CAS No.: 22439-61-8 Molecular Formula: C12H7BrS Formula Weight: 263.15 Melting Point: 123-129° lit Beilstein Reference: 142203 MDL No.: MFCD00089285

Formula: C₁₂H₇BrS
Peso molecolare: 263,15 g/mol
Punto di fusione: 123…129 °C (lett.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00089285
CAS Number: 22439-61-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR