Stampa…

5-Methylpicolinonitrile 97+%

Fornitore: Thermo Fisher Scientific
Pericolo

H64539.06 H64539.14
H64539.06EA 93.6 EUR
H64539.06 H64539.14
5-Methylpicolinonitrile 97+%
5-Methylpicolinonitrile

Purity: 97+% CAS No.: 1620-77-5 Molecular Formula: C7H6N2 Formula Weight: 118.14 Physical Form: Solid Melting Point: 72-75° Beilstein Reference: 111125 UN No.: UN3439 MDL No.: MFCD06200830

Formula: C7H6N2
Punto di fusione: 72…75 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06200830
CAS Number: 1620-77-5
EINECS: 216-589-4
ADR: 6.1,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR