N-(4-Oxocyclohexyl)acetamide 98%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
4-Acetamidocyclohexanone
Purity: 98%
CAS No.: 27514-08-5
Molecular Formula: C8H13NO2
Formula Weight: 155.20
Physical Form: Solid
Appearance: Off-white
Melting Point: 136-140°
Beilstein Reference: 2803083
MDL No.: MFCD03703462
Formula:
C₈H₁₃NO₂ Peso molecolare: 155,20 g/mol Punto di fusione: 136…140 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03703462 CAS Number: 27514-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...