Stampa…

2-Bromo-5-chlorobenzaldehyde 97%

Fornitore: Thermo Scientific
Attenzione

H64330.06 H64330.14 H64330.22
H64330.06EA 34.1 EUR
H64330.06 H64330.14 H64330.22
2-Bromo-5-chlorobenzaldehyde 97%
2-Bromo-5-chlorobenzaldehyde

Purity: 97% CAS No.: 174265-12-4 Molecular Formula: C7H4BrClO Formula Weight: 219.46 Physical Form: Crystalline Appearance: Pale yellow Melting Point: 64-68° lit. Beilstein Reference: 10423891 MDL No.: MFCD00462870

Formula: C₇H₄BrClO
Punto di fusione: 64…68 °C (lett.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00462870
CAS Number: 174265-12-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR