2-Amino-4-fluoropyridine 98%
Fornitore: Thermo Scientific
|
Attenzione
|
H64233.03
H64233.06
H64233.03EA
139
EUR
H64233.03
H64233.06
2-Amino-4-fluoropyridine 98%
2-Amino-4-fluoropyridine
Purity: 98%
CAS No.: 944401-77-8
Molecular Formula: C5H5FN2
Formula Weight: 112.04
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Pale yellow
Beilstein Reference: 15519014
MDL No.: MFCD09260906
Formula:
C₅H₅FN₂ Peso molecolare: 112,04 g/mol |
MDL Number:
MFCD09260906 CAS Number: 944401-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...