Tri-p-tolylamine 98%
Fornitore: Thermo Scientific
Synonyms:
4,4',4''-Trimethyltriphenylamine
Purity: 98%
CAS No.: 1159-53-1
Molecular Formula: C21H21N
Formula Weight: 287.41
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White to off-white
Melting Point: 114-118° lit.
Beilstein Reference: 2215309
MDL No.: MFCD00674043
Formula:
C₂₁H₂₁N Peso molecolare: 287,41 g/mol Punto di fusione: 114…118 °C (lett.) |
MDL Number:
MFCD00674043 CAS Number: 1159-53-1 EINECS: 214-595-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...