4-(Boc-amino)indole-3-carboxaldehyde 97%
Fornitore: Thermo Scientific
|
Attenzione
|
H63587.MD
H63587.03
H63587.MDEA
125
EUR
H63587.MD
H63587.03
4-(Boc-amino)indole-3-carboxaldehyde 97%
4-(Boc-amino)indole-3-carboxaldehyde
Purity: 97%
CAS No.: 885266-77-3
Molecular Formula: C14H16N2O3
Formula Weight: 260.29
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD04973990
Formula:
C₁₄H₁₆N₂O₃ |
MDL Number:
MFCD04973990 CAS Number: 885266-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...