Stampa…

tert-Butyl-2-(methylamino)acetate hydrochloride 97%

Fornitore: Thermo Scientific

Synonyms: Sar-OtBu·HCl, tert-Butyl sarcosinate hydrochloride, Sarcosine tert-butyl ester hydrochloride, H-Sar-OtBu·HCl, tert-Butyl 2-(methylamino)acetate hydrochloride, H-Sar-OtBu.HCl

H63176.06 H63176.14
H63176.06EA 79 EUR
H63176.06 H63176.14
tert-Butyl-2-(methylamino)acetate hydrochloride 97%
tert-Butyl-2-(methylamino)acetate hydrochloride

Purity: 97% CAS No.: 5616-81-9 Molecular Formula: C7H15NO2.HCl Formula Weight: 181.66 Physical Form: Powder Appearance: White Melting Point: 137-141° MDL No.: MFCD00080936

Formula: C₇H₁₆ClNO₂
Peso molecolare: 181,660 g/mol
Punto di fusione: 137…141 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00080936
CAS Number: 5616-81-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR