Stampa…

Fmoc-6-Ahx-OH 98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: Fmoc-ε-aminocaproic acid, Fmoc-ε-Acp-OH, 6-(Fmoc-amino)hexanoic acid

H63148.03 H63148.06 H63148.14
H63148.03EA 38.4 EUR
H63148.03 H63148.06 H63148.14
Fmoc-6-Ahx-OH 98%
Fmoc-6-Ahx-OH

Purity: 98% CAS No.: 88574-06-5 Molecular Formula: C21H23NO4 Formula Weight: 353.41 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Beilstein Reference: 5883220 MDL No.: MFCD00077045

Formula: C₂₁H₂₃NO₄
Peso molecolare: 353,418 g/mol
MDL Number: MFCD00077045
CAS Number: 88574-06-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR