Boc-D-Glu-OtBu 95%
Fornitore: Thermo Scientific
Synonyms:
N-Boc-D-glutamic acid 1-tert-butyl ester
Purity: 95%
CAS No.: 73872-71-6
Molecular Formula: C14H25NO6
Formula Weight: 303.35
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale brown
MDL No.: MFCD00076926
Formula:
C₁₄H₂₅NO₆ |
MDL Number:
MFCD00076926 CAS Number: 73872-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...