7-Chloroisatin 97%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
7-Chloroindoline-2,3-dione, 7-Chloro-1H-indole-2,3-dione, 7-Cloroindoline-2,3-dione
Purity: 97%
CAS No.: 7477-63-6
Molecular Formula: C8H4ClNO2
Formula Weight: 181.58
Physical Form: Solid
Appearance: Orange
Melting Point: 187-191°
MDL No.: MFCD00022796
Formula:
C₈H₄ClNO₂ Peso molecolare: 181,58 g/mol Punto di fusione: 187…191 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00022796 CAS Number: 7477-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...