Stampa…

4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline 97%

Fornitore: Thermo Scientific
Attenzione

Synonyms: N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-iodoquinazolin-4-amine

H54229.MD H54229.03
H54229.MDEA 251 EUR
H54229.MD H54229.03
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline 97%
4-[3-Chloro-4-(3-fluorobenzyloxy)phenylamino]-6-iodoquinazoline

Purity: 97% CAS No.: 231278-20-9 Molecular Formula: C21H14ClFIN3O Formula Weight: 505.72 MDL No.: MFCD09998827

Formula: C₂₁H₁₄ClFIN₃O
Peso molecolare: 505,72 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09998827
CAS Number: 231278-20-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR