N,N-Dimethylisobutyramide 99%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 99%
CAS No.: 21678-37-5
Molecular Formula: C6H13NO
Formula Weight: 115.18
Flash Point: 154°(309°F)
Refractive Index: 1.439
Boiling Point: 175-176°/44mm
Beilstein Reference: 1742178
MDL No.: MFCD00075378
Formula:
C₆H₁₃NO Peso molecolare: 115.18 g/mol Punto di ebollizione: 175…176 °C (44 mmHg) Densità: 0.892 Flash Pt: 154 °C (309 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00075378 CAS Number: 21678-37-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...