Stampa…

(2-Aminoethoxy)acetic acid 98%

Fornitore: Thermo Scientific

H52289.03 H52289.06
H52289.03EA 221 EUR
H52289.03 H52289.06
(2-Aminoethoxy)acetic acid 98%
(2-Aminoethoxy)acetic acid

Purity: 98% CAS No.: 10366-71-9 Molecular Formula: C4H9NO3 Formula Weight: 119.12 Melting Point: 177-179° MDL No.: MFCD09865417

Formula: C₄H₉NO₃
Peso molecolare: 119,12 g/mol
Punto di fusione: 177…179 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09865417
CAS Number: 10366-71-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR