Stampa…
3-Chlorophenylmagnesium bromide 1 M in MeTHF (2-methyltetrahydrofuran)
Fornitore: Thermo Fisher Scientific
Pericolo
H51161.AD H51161.AE
H51161.ADEA 182 EUR
H51161.AD H51161.AE
3-Chlorophenylmagnesium bromide 1 M in MeTHF (2-methyltetrahydrofuran)
3-Chlorophenylmagnesium bromide
CAS No.: 36229-42-2 Molecular Formula: C6H4BrClMg Formula Weight: 215.76 UN No.: UN2924 MDL No.: MFCD00672003
Formula: C₆H₄BrClMg
Peso molecolare: 215.757 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00672003
CAS Number: 36229-42-2
UN: 2924
ADR: 3 II