Stampa…

1-(2-Fluorophenyl)-2-propylamine 97%

Fornitore: Thermo Scientific
Attenzione

H50999.03 H50999.MD
H50999.03EA 252 EUR
H50999.03 H50999.MD
1-(2-Fluorophenyl)-2-propylamine 97%
1-(2-Fluorophenyl)-2-propylamine

Purity: 97% CAS No.: 1716-60-5 Molecular Formula: C9H12FN Formula Weight: 153.20 MDL No.: MFCD01708100

Formula: C₉H₁₂FN
Peso molecolare: 153,20 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01708100
CAS Number: 1716-60-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR