Stampa…

3,5-Dimethyl-1H-pyrrole-2-carbaldehyde 97%

Fornitore: Thermo Scientific
Attenzione

H50478.03 H50478.MD
H50478.03EA 212 EUR
H50478.03 H50478.MD
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde 97%
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde

Purity: 97% CAS No.: 2199-58-8 Molecular Formula: C7H9NO Formula Weight: 123.16 MDL No.: MFCD00111522

Formula: C7H9NO
Peso molecolare: 123,16 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00111522
CAS Number: 2199-58-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR