Isopropyl glycolate 96%
Fornitore: Thermo Scientific
|
Attenzione
|
H50334.03
H50334.06
H50334.03EA
96.1
EUR
H50334.03
H50334.06
Isopropyl glycolate 96%
Isopropyl glycolate
Purity: 96%
CAS No.: 623-61-0
Molecular Formula: HOCH2CO2CH(CH3)2
Formula Weight: 118.13
MDL No.: MFCD00046747
Formula:
HOCH₂CO₂CH(CH₃)₂ Peso molecolare: 118,13 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00046747 CAS Number: 623-61-0 EINECS: 210-804-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...