Stampa…

3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] 96%

Fornitore: Thermo Scientific
Attenzione

Synonyms: DiSC3(5), 3-Propyl-2-[5-[3-propyl-2(3H)-benzothiazolylidene]-1,3-pentadienyl]benzothiazolium iodide, 5-(3-Propylbenzothiazol-2-ylidene)-1-(3-propylbenzothiazolium-2-yl)-1,3-pentadiene odide, 3,3'-Dipropylthiadicarbocyanine iodide

H34209.MD H34209.03
H34209.MDEA 106 EUR
H34209.MD H34209.03
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] 96%
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)]

Purity: 96% CAS No.: 53213-94-8 Molecular Formula: C25H27IN2S2 Formula Weight: 546.54 Melting Point: 250° MDL No.: MFCD00075033

Formula: C₂₅H₂₇IN₂S₂
Peso molecolare: 546,53 g/mol
Punto di fusione: >250 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00075033
CAS Number: 53213-94-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR