Stampa…

Trimethylolpropane tris[3-(2-methylaziridin-1-yl)propionate]

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: 2-Ethyl-2-(((3-(2-methylaziridin-1-yl)propanoyl)oxy)methyl)propane-1,3-diyl bis(3-(2-methylaziridin-1-yl)propanoate)

H33949.03 H33949.06
H33949.03EA 57.5 EUR
H33949.03 H33949.06
Trimethylolpropane tris[3-(2-methylaziridin-1-yl)propionate]
Trimethylolpropane tris[3-(2-methylaziridin-1-yl)propionate]

CAS No.: 64265-57-2 Molecular Formula: C24H41N3O6 Formula Weight: 467.61 Melting Point: -30° MDL No.: MFCD00192542

Formula: C₂₄H₄₁N₃O₆
Punto di fusione: –30 °C
Densità: 1,08
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00192542
CAS Number: 64265-57-2
EINECS: 264-763-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR