Stampa…

N-Formylglycine 98%

Fornitore: Thermo Scientific

Synonyms: For-Gly-OH

H31053.03 H31053.06
H31053.03EA 41.2 EUR
H31053.03 H31053.06
N-Formylglycine 98%
N-Formylglycine

Purity: 98% CAS No.: 2491-15-8 Molecular Formula: C3H5NO3 Formula Weight: 103.08 Storage Temperature: 2°C to 8°C Melting Point: 147-151° MDL No.: MFCD00037360

Formula: C₃H₅NO₃
Peso molecolare: 103,08 g/mol
Punto di fusione: 147…151 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00037360
CAS Number: 2491-15-8
EINECS: 219-649-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR